Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064747.1 | XP_017920236.1 | 2 - 3 | 206 | 88 | |
GenBank mRNA | JO630623.2 | - | 4 - 5 | 1545 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |