Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685754.3 | XP_005685811.2 | 7 - 8 | 947 | 92 | |
GenBank mRNA | JT710956.1 | - | 7 - 8 | 711 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |