Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054789.1 | XP_017910278.1 | 3 - 4 | 572 | 92 | |
GenBank mRNA | KA334015.1 | - | 2 - 3 | 416 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |