Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INO80C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696979.3 | XP_005697036.1 | 5 | 854 | 110 | |
XM_013974312.2 | XP_013829766.1 | 5 | 719 | 110 | ||
XM_013974314.2 | XP_013829768.1 | 5 | 790 | 110 | ||
XM_013974315.2 | XP_013829769.1 | 5 | 867 | 110 | ||
XM_018039706.1 | XP_017895195.1 | 6 | 943 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |