Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172953.1 | NP_001166424.1 | 6 - 7 | 874 | 91 | |
GenBank mRNA | D21232.1 | - | 6 - 7 | 874 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Cavia porcellus Probe spans exons |