Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172918.2 | NP_001166389.1 | 5 - 6 | 685 | 123 | |
GenBank mRNA | D84448.1 | - | 5 - 6 | 686 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Cavia porcellus Probe spans exons |