Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172926.1 | NP_001166397.1 | 1 - 2 | 113 | 89 | |
GenBank mRNA | L04985.1 | - | 1 - 2 | 113 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Cavia porcellus Probe spans exons |