Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172951.1 | NP_001166422.1 | 9 - 10 | 812 | 78 | |
GenBank mRNA | AB060340.1 | - | 1 | 746 | 78 | |
EU862201.1 | - | 9 - 10 | 812 | 78 | ||
U51572.1 | - | 1 | 259 | 78 | ||
U76741.1 | - | 2 - 3 | 152 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |