Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173073.1 | NP_001166544.1 | 1 - 2 | 96 | 57 | |
XM_005004205.3 | XP_005004262.1 | 2 - 3 | 225 | 57 | ||
XM_005004206.3 | XP_005004263.1 | 2 - 3 | 185 | 57 | ||
XM_005004207.3 | XP_005004264.1 | 2 - 3 | 225 | 57 | ||
GenBank mRNA | EU306867.1 | - | 1 - 2 | 96 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Cavia porcellus Probe spans exons |