Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ku80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135923.3 | NP_609767.2 | 4 - 5 | 565 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |