Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078983.4 | NP_523707.1 | 2 - 3 | 1210 | 60 | |
NM_165867.2 | NP_725107.1 | 2 - 3 | 1016 | 60 | ||
GenBank mRNA | AF172635.1 | - | 2 - 3 | 1179 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |