Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nemy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169657.2 | NP_001163128.1 | 3 - 4 | 571 | 66 | |
NM_001299429.1 | NP_001286358.1 | 2 - 3 | 1038 | 66 | ||
NM_001299430.1 | NP_001286359.1 | 2 - 3 | 713 | 66 | ||
NM_136964.4 | NP_610808.1 | 2 - 3 | 1038 | 66 | ||
NM_165934.3 | NP_725208.1 | 3 - 4 | 716 | 66 | ||
NM_165935.3 | NP_725209.1 | 3 - 4 | 526 | 66 | ||
NM_165936.3 | NP_725210.1 | 2 - 3 | 323 | 66 | ||
NM_165937.3 | NP_725211.3 | 2 - 3 | 521 | 66 | ||
GenBank mRNA | AY060294.1 | - | 2 - 3 | 1024 | 66 | |
BT003222.1 | - | 3 - 4 | 714 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |