Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nemy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169657.2 | NP_001163128.1 | 5 - 6 | 959 | 53 | |
NM_136964.4 | NP_610808.1 | 4 - 5 | 1426 | 53 | ||
NM_165934.3 | NP_725208.1 | 5 - 6 | 1104 | 53 | ||
NM_165935.3 | NP_725209.1 | 5 - 6 | 914 | 53 | ||
NM_165936.3 | NP_725210.1 | 4 - 5 | 711 | 53 | ||
GenBank mRNA | AY060294.1 | - | 4 - 5 | 1412 | 53 | |
BT003222.1 | - | 5 - 6 | 1102 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |