Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nemy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169657.2 | NP_001163128.1 | 2 - 3 | 349 | 67 | |
NM_001299429.1 | NP_001286358.1 | 1 - 2 | 816 | 67 | ||
NM_136964.4 | NP_610808.1 | 1 - 2 | 816 | 67 | ||
NM_165935.3 | NP_725209.1 | 2 - 3 | 304 | 67 | ||
GenBank mRNA | AY060294.1 | - | 1 - 2 | 802 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |