Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cad74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259885.1 | NP_001246814.1 | 5 - 6 | 2884 | 85 | |
NM_140716.2 | NP_648973.1 | 5 - 6 | 2598 | 85 | ||
NM_168724.2 | NP_730261.1 | 5 - 6 | 2613 | 85 | ||
GenBank mRNA | BT150474.1 | - | 5 - 6 | 2612 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |