Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014503.2 | NP_001014503.1 | 1 - 2 | 449 | 77 | |
NM_001169610.2 | NP_001163081.1 | 2 - 3 | 442 | 77 | ||
NM_001169611.1 | NP_001163082.1 | 2 - 3 | 466 | 77 | ||
NM_001169612.2 | NP_001163083.1 | 3 - 4 | 508 | 77 | ||
NM_001259270.2 | NP_001246199.1 | 2 - 3 | 442 | 77 | ||
NM_001259271.2 | NP_001246200.1 | 2 - 3 | 442 | 77 | ||
NM_001273865.1 | NP_001260794.1 | 1 - 2 | 449 | 77 | ||
NM_001273866.2 | NP_001260795.1 | 2 - 3 | 442 | 77 | ||
NM_001316409.1 | NP_001303338.1 | 2 - 3 | 442 | 77 | ||
NM_136504.5 | NP_610348.3 | 2 - 3 | 442 | 77 | ||
NM_165586.5 | NP_724643.2 | 2 - 3 | 511 | 77 | ||
NM_206056.5 | NP_995778.2 | 2 - 3 | 797 | 77 | ||
GenBank mRNA | AF276964.1 | - | 1 - 2 | 449 | 77 | |
AY069682.1 | - | 2 - 3 | 466 | 77 | ||
BT100315.1 | - | 1 - 2 | 415 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |