Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297991.1 | NP_001284920.1 | 5 - 6 | 854 | 61 | |
NM_001297992.1 | NP_001284921.1 | 4 - 5 | 805 | 61 | ||
NM_132057.3 | NP_572285.2 | 4 - 5 | 992 | 61 | ||
GenBank mRNA | BT044143.1 | - | 4 - 5 | 991 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |