Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297991.1 | NP_001284920.1 | 7 - 8 | 1323 | 86 | |
NM_001297992.1 | NP_001284921.1 | 6 - 7 | 1274 | 86 | ||
NM_132057.3 | NP_572285.2 | 6 - 7 | 1461 | 86 | ||
GenBank mRNA | BT044143.1 | - | 6 - 7 | 1460 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |