Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299095.1 | NP_001286024.1 | 5 - 6 | 1053 | 93 | |
NM_136017.3 | NP_609861.1 | 5 - 6 | 1053 | 93 | ||
GenBank mRNA | BT004502.1 | - | 5 - 6 | 1026 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |