Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Ea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080273.3 | NP_525012.2 | 5 - 6 | 764 | 60 | |
NM_144389.2 | NP_652646.1 | 5 - 6 | 759 | 60 | ||
NM_165502.2 | NP_724524.1 | 5 - 6 | 753 | 60 | ||
GenBank mRNA | AF274014.1 | - | 5 - 6 | 751 | 60 | |
AY069049.1 | - | 5 - 6 | 751 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |