Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259396.2 | NP_001246325.1 | 4 - 5 | 1110 | 69 | |
NM_058126.5 | NP_477474.1 | 3 - 4 | 1275 | 69 | ||
NM_166065.3 | NP_725412.2 | 4 - 5 | 1162 | 69 | ||
GenBank mRNA | AF145309.1 | - | 3 - 4 | 1037 | 69 | |
AF160939.1 | - | 3 - 4 | 1245 | 69 | ||
BT089015.1 | - | 4 - 5 | 1142 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |