Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274064.2 | NP_001260993.1 | 3 - 4 | 872 | 68 | |
NM_001382195.1 | NP_001369087.1 | 3 - 4 | 872 | 68 | ||
NM_137190.4 | NP_611034.2 | 2 - 3 | 867 | 68 | ||
NM_166093.3 | NP_725458.1 | 3 - 4 | 872 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |