Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299540.1 | NP_001286469.1 | 7 - 8 | 3733 | 65 | |
NM_137220.3 | NP_611064.2 | 7 - 8 | 3736 | 65 | ||
NM_166121.2 | NP_725499.2 | 6 - 7 | 4243 | 65 | ||
NM_176181.2 | NP_788361.1 | 7 - 8 | 3949 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |