Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166134.3 | NP_725517.2 | 1 - 2 | 127 | 72 | |
GenBank mRNA | BT024331.1 | - | 409 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |