Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137242.3 | NP_611086.1 | 2 - 3 | 311 | 61 | |
NM_166148.2 | NP_725534.1 | 3 - 4 | 310 | 61 | ||
GenBank mRNA | AY061313.1 | - | 2 - 3 | 284 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |