Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043154.4 | NP_001036619.1 | 2 - 3 | 937 | 112 | |
NM_001170022.3 | NP_001163493.1 | 2 - 3 | 937 | 112 | ||
NM_001259944.2 | NP_001246873.1 | 3 - 4 | 1049 | 112 | ||
NM_001259945.2 | NP_001246874.1 | 2 - 3 | 937 | 112 | ||
NM_001259946.2 | NP_001246875.1 | 2 - 3 | 937 | 112 | ||
NM_001259947.1 | NP_001246876.1 | 3 - 4 | 1049 | 112 | ||
NM_001275268.1 | NP_001262197.1 | 3 - 4 | 1049 | 112 | ||
NM_001275269.1 | NP_001262198.1 | 3 - 4 | 996 | 112 | ||
NM_001316446.1 | NP_001303375.1 | 2 - 3 | 937 | 112 | ||
NM_079484.6 | NP_524208.1 | 2 - 3 | 937 | 112 | ||
NM_168946.4 | NP_730700.2 | 3 - 4 | 1049 | 112 | ||
NM_206423.5 | NP_996145.1 | 2 - 3 | 937 | 112 | ||
GenBank mRNA | AY060403.1 | - | 2 - 3 | 885 | 112 | |
BT030417.1 | - | 2 - 3 | 873 | 112 | ||
X99340.1 | - | 1692 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster |