Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mael | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316461.1 | NP_001303390.1 | 3 - 4 | 957 | 63 | |
NM_079493.4 | NP_524217.1 | 3 - 4 | 573 | 63 | ||
NM_168958.3 | NP_730739.1 | 2 - 3 | 644 | 63 | ||
NM_176388.3 | NP_788566.1 | 2 - 3 | 653 | 63 | ||
GenBank mRNA | AY119598.1 | - | 2 - 3 | 643 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |