Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc68E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206342.2 | NP_996064.1 | 5358 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |