Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274827.1 | NP_001261756.1 | 11 - 12 | 2937 | 70 | |
NM_001274828.1 | NP_001261757.1 | 11 - 12 | 2111 | 70 | ||
NM_001274829.1 | NP_001261758.1 | 11 - 12 | 2111 | 70 | ||
NM_001274830.2 | NP_001261759.1 | 11 - 12 | 2166 | 70 | ||
NM_140315.4 | NP_648572.1 | 11 - 12 | 2111 | 70 | ||
NM_206344.2 | NP_996066.1 | 11 - 12 | 2937 | 70 | ||
GenBank mRNA | AY094735.1 | - | 11 - 12 | 2082 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |