Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042794.4 | NP_001036259.2 | 9 - 10 | 2552 | 50 | |
NM_001103400.2 | NP_001096870.1 | 9 - 10 | 2158 | 50 | ||
NM_001272264.1 | NP_001259193.1 | 10 - 11 | 3754 | 50 | ||
NM_057366.5 | NP_476714.1 | 9 - 10 | 2184 | 50 | ||
NM_206613.3 | NP_996336.1 | 11 - 12 | 2473 | 50 | ||
GenBank mRNA | X70863.1 | - | 2143 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |