Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080344.3 | NP_525083.2 | 1 - 2 | 350 | 76 | |
NM_167232.2 | NP_727427.1 | 2 - 3 | 379 | 76 | ||
NM_206677.2 | NP_996400.1 | 2 - 3 | 362 | 76 | ||
GenBank mRNA | AY051757.1 | - | 2 - 3 | 375 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |