Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pino | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272925.1 | NP_001259854.1 | 3 - 4 | 589 | 63 | |
NM_134724.3 | NP_608568.1 | 2 - 3 | 538 | 63 | ||
NM_164405.2 | NP_722679.1 | 3 - 4 | 439 | 63 | ||
GenBank mRNA | AY069447.1 | - | 2 - 3 | 515 | 63 | |
BT133460.1 | - | 3 - 4 | 425 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |