Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144444.2 | NP_001137916.1 | 5 - 6 | 1882 | 82 | |
NM_001144445.2 | NP_001137917.1 | 4 - 5 | 1352 | 82 | ||
NM_001144446.2 | NP_001137918.1 | 5 - 6 | 1828 | 82 | ||
NM_001144447.2 | NP_001137919.1 | 5 - 6 | 1419 | 82 | ||
NM_001144448.2 | NP_001137920.1 | 4 - 5 | 1465 | 82 | ||
NM_001144449.2 | NP_001137921.1 | 4 - 5 | 1339 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |