Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140023.3 | NP_648280.1 | 1 - 2 | 214 | 53 | |
GenBank mRNA | AY071335.1 | - | 1 - 2 | 212 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |