Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: defl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140095.4 | NP_648352.2 | 1 - 2 | 1591 | 65 | |
GenBank mRNA | BT044201.1 | - | 1 - 2 | 1504 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |