Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JIL-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274768.1 | NP_001261697.1 | 4 - 5 | 2004 | 62 | |
NM_140175.3 | NP_648432.1 | 4 - 5 | 2004 | 62 | ||
NM_168439.2 | NP_729661.1 | 3 - 4 | 1166 | 62 | ||
GenBank mRNA | AF142061.1 | - | 4 - 5 | 1959 | 62 | |
BT044484.1 | - | 3 - 4 | 1158 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |