Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274823.1 | NP_001261752.1 | 1 | 1168 | 155 | |
NM_140307.4 | NP_648564.1 | 1 | 1168 | 155 | ||
GenBank mRNA | AY071047.1 | - | 1 | 1153 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |