Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274840.1 | NP_001261769.1 | 6 - 7 | 2338 | 116 | |
NM_001274841.1 | NP_001261770.1 | 6 - 7 | 2350 | 116 | ||
NM_001274842.1 | NP_001261771.1 | 6 - 7 | 2320 | 116 | ||
NM_001274843.1 | NP_001261772.1 | 6 - 7 | 2308 | 116 | ||
NM_001274844.1 | NP_001261773.1 | 6 - 7 | 2341 | 116 | ||
NM_001274845.1 | NP_001261774.1 | 6 - 7 | 2311 | 116 | ||
NM_001274846.1 | NP_001261775.1 | 6 - 7 | 2353 | 116 | ||
NM_001274847.1 | NP_001261776.1 | 6 - 7 | 2323 | 116 | ||
NM_140328.5 | NP_648585.3 | 6 - 7 | 2338 | 116 | ||
GenBank mRNA | AY069066.1 | - | 6 - 7 | 2320 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |