Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258983.2 | NP_001245912.1 | 5 - 6 | 1200 | 92 | |
NM_001273193.1 | NP_001260122.1 | 8 - 9 | 1477 | 92 | ||
NM_164682.3 | NP_723156.1 | 6 - 7 | 941 | 92 | ||
NM_164683.3 | NP_723157.1 | 6 - 7 | 1305 | 92 | ||
GenBank mRNA | AF202774.1 | - | 6 - 7 | 863 | 92 | |
BT001751.1 | - | 6 - 7 | 940 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |