Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078782.3 | NP_523506.1 | 2 - 3 | 1536 | 86 | |
NM_164761.2 | NP_723298.1 | 2 - 3 | 1536 | 86 | ||
NM_164762.2 | NP_723299.2 | 2 - 3 | 1536 | 86 | ||
NM_164763.2 | NP_723300.2 | 2 - 3 | 1536 | 86 | ||
NM_164764.2 | NP_723301.1 | 2 - 3 | 1536 | 86 | ||
GenBank mRNA | AJ269539.1 | - | 2 - 3 | 1520 | 86 | |
AY122084.1 | - | 2 - 3 | 1520 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |