Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042881.5 | NP_001036346.3 | 13 - 14 | 3949 | 97 | |
NM_001169436.3 | NP_001162907.1 | 12 - 13 | 3727 | 97 | ||
NM_001201789.2 | NP_001188718.1 | 13 - 14 | 3784 | 97 | ||
NM_001201790.2 | NP_001188719.1 | 12 - 13 | 3727 | 97 | ||
NM_001273297.1 | NP_001260226.1 | 14 - 15 | 4006 | 97 | ||
NM_001273298.1 | NP_001260227.1 | 13 - 14 | 3793 | 97 | ||
NM_001273299.1 | NP_001260228.1 | 12 - 13 | 3727 | 97 | ||
NM_001273300.2 | NP_001260229.2 | 12 - 13 | 3727 | 97 | ||
NM_001316385.1 | NP_001303314.1 | 12 - 13 | 3727 | 97 | ||
NM_135344.6 | NP_609188.4 | 13 - 14 | 3723 | 97 | ||
NM_164795.6 | NP_723354.3 | 13 - 14 | 3949 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |