Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042881.5 | NP_001036346.3 | 15 - 16 | 4911 | 63 | |
NM_001169436.3 | NP_001162907.1 | 14 - 15 | 4689 | 63 | ||
NM_001201789.2 | NP_001188718.1 | 15 - 16 | 4746 | 63 | ||
NM_001201790.2 | NP_001188719.1 | 14 - 15 | 4689 | 63 | ||
NM_001273297.1 | NP_001260226.1 | 16 - 17 | 4968 | 63 | ||
NM_001273299.1 | NP_001260228.1 | 14 - 15 | 4689 | 63 | ||
NM_135344.6 | NP_609188.4 | 15 - 16 | 4685 | 63 | ||
NM_164795.6 | NP_723354.3 | 15 - 16 | 4911 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |