Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: salr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259054.2 | NP_001245983.1 | 4 - 5 | 3975 | 57 | |
NM_078824.3 | NP_523548.1 | 4 - 5 | 3167 | 57 | ||
NM_164965.2 | NP_723669.1 | 4 - 5 | 3433 | 57 | ||
GenBank mRNA | BT150428.1 | - | 4 - 5 | 3404 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |