Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgant5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042873.2 | NP_001036338.1 | 7 - 8 | 2246 | 71 | |
NM_135062.4 | NP_608906.2 | 8 - 9 | 2270 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |