Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TpnC25D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298690.1 | NP_001285619.1 | 2 - 3 | 976 | 70 | |
NM_001298691.1 | NP_001285620.1 | 2 - 3 | 557 | 70 | ||
NM_135071.4 | NP_608915.2 | 2 - 3 | 514 | 70 | ||
GenBank mRNA | AY113496.1 | - | 2 - 3 | 508 | 70 | |
AY283601.1 | - | 2 - 3 | 508 | 70 | ||
KX531911.1 | - | 1 - 2 | 417 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |