Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140385.4 | NP_648642.1 | 6 - 7 | 1822 | 67 | |
NM_168543.2 | NP_729891.1 | 6 - 7 | 1757 | 67 | ||
GenBank mRNA | AY061340.1 | - | 6 - 7 | 1803 | 67 | |
BT099796.1 | - | 6 - 7 | 1757 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |