Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eig71Ej | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140539.1 | NP_648796.1 | 1 - 2 | 243 | 75 | |
GenBank mRNA | KX531950.1 | - | 1 - 2 | 231 | 75 | |
U24574.1 | - | 1 - 2 | 243 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |