Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hip14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274981.1 | NP_001261910.1 | 2 - 3 | 1630 | 62 | |
NM_001274982.1 | NP_001261911.1 | 2 - 3 | 1630 | 62 | ||
NM_140567.3 | NP_648824.1 | 2 - 3 | 1630 | 62 | ||
GenBank mRNA | AY094779.1 | - | 2 - 3 | 1600 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |