Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140809.3 | NP_649066.1 | 5 - 6 | 1469 | 78 | |
GenBank mRNA | AF124502.1 | - | 5 - 6 | 1437 | 78 | |
AY061116.1 | - | 5 - 6 | 1436 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |