Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir76b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140919.2 | NP_649176.1 | 4 - 5 | 1397 | 96 | |
GenBank mRNA | BT022767.1 | - | 4 - 5 | 1388 | 96 | |
BT022775.1 | - | 4 - 5 | 1397 | 96 | ||
HQ600589.1 | - | 4 - 5 | 1300 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |